Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-17RB Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10402G02
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10402G02 25 Tests
FAB10402G10 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB10402G02 Supplier R&D Systems Supplier No. FAB10402G025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 1 publication

IL-17RB Monoclonal specifically detects IL-17RB in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RB
Applications Flow Cytometry
Classification Monoclonal
Clone 752101
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9JIP3
Gene Alias CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor
Gene Symbols IL17RB
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RB Arg18-Gly286 Accession # Q9JIP3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55540
Test Specificity Detects mouse IL-17 RB in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 RB, rhIL-17 R, or recombinant mouse IL-17 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.