Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-17RC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2270
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2270 Supplier R&D Systems Supplier No. AF2270
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

IL-17RC Polyclonal specifically detects IL-17RC in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen IL-17RC
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.6-3.6 ug/mL, CyTOF-ready
Gene Accession No. AAH04759
Gene Alias FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14
Gene Symbols IL17RC
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RC Leu21-Trp465 Accession # AAH04759
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84818
Test Specificity Detects mouse IL-17 RC in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IL-17 RC is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-17 RD and rmIL-17B R is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.