Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RC APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB2270A100UG
This item is not returnable.
View return policy
Description
IL-17RC Polyclonal specifically detects IL-17RC in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-17RC | |
Polyclonal | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH04759 | |
IL17RC | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RC, Leu21-Trp465, Accession No. AAH04759 | |
100 μg | |
84818.0 | |
Mouse | |
IgG |
Flow Cytometry | |
APC | |
Supplied in a saline solution containing BSA and sodium azide | |
FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
Goat | |
Antigen Affinity-purified | |
Primary | |
Detects mouse IL-17 RC in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IL-17 RC is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-17 RD and rmIL-17B R is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction