Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RD/SEF Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB2276S100UG
This item is not returnable.
View return policy
Description
IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-17RD/SEF | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
FLJ35755, IL-17 RD, IL17RD, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLM, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, SEF, Sef homolog, SEFhSef | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RD/SEF, Gly28-Arg299, Accession # Q8JZL1 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
400210 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8JZL1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54756 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction