Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RE Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7997G100UG
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-17RE | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E | |
Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE Met1-His414 Accession # Q8BH06 | |
RUO | |
132014 | |
Mouse | |
IgG1 |
Flow Cytometry | |
944904 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8BH06 | |
Rat | |
100 μg | |
Primary | |
Detects mouse IL-17 RE in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction