Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-18/IL-1F4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. D0473
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D0473 Supplier R&D Systems Supplier No. D0473
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Mouse samples. It is validated for Immunoprecipitation, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-18/IL-1F4
Applications Immunoprecipitation, ELISA
Classification Monoclonal
Clone 74
Conjugate Unconjugated
Dilution Immunoprecipitation, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation 100 μg IgG in 100 μL volume of PBS containing 50% glycerol, pH 7.2. No preservative is contained. with No Preservative
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Gene Symbols IL18
Host Species Rat
Immunogen Mouse IL-18/IL-1F4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity This antibody reacts with mouse IL-18 on Immunoprecipitation and ELISA.
Target Species Mouse
Content And Storage This antibody solution is stable for one year from the date of purchase when stored at -20 degreesC.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.