Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-18 R alpha/IL-1 R5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1216
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IL-18R alpha/IL-1 R5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q61098 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8809 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
112614 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Il18rap | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5 Ser19-Gly326 Accession # Q61098 | |
500 μg | |
Primary | |
Detects mouse IL-18 R alpha/IL-1 R5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, recombinant human (rh) IL-1 R3, rhIL-1 R4, recombinant rat (rr) IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction