Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-18 R alpha/IL-1 R5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB12161500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB12161500 500 μg
MAB12161SP 25 μg
MAB12161100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB12161500 Supplier R&D Systems Supplier No. MAB12161500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 112624
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61098
Gene Symbols Il18rap
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha Ser19-Gly326 Accession # Q61098
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Test Specificity Detects mouse IL-18 R in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, rhIL-1 R3, recombinant human (rh) IL-1 R4, rhIL-1 R5, recombinant rat IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.