Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-18 R alpha/IL-1 R5 Fluorescein-conjugated Antibody, R&D Systems™

Catalog No. FAB1216F025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1216F025 25 Tests
FAB1216F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1216F025 Supplier R&D Systems Supplier No. FAB1216F025
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications Flow Cytometry
Classification Monoclonal
Clone 112614
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q61098
Gene Symbols Il18rap
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5 Ser19-Gly326 Accession # Q61098
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Test Specificity Detects mouse IL-18 R alpha/IL-1 R5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, recombinant human (rh) IL-1 R3, rhIL-1 R4, recombinant rat (rr) IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.