Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Goat Polyclonal Antibody
$227.50 - $549.50
Specifications
Antigen | IL-1Rrp2/IL-1R6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-1Rrp2/IL-1R6 Polyclonal specifically detects IL-1Rrp2/IL-1R6 in Mouse samples. It is validated for Western Blot.Specifications
IL-1Rrp2/IL-1R6 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
Il1rl2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9ERS7 | |
8808 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 Rrp2/IL-1 R6 Asp22-Arg338 (Ala158Val) Accession # Q9ERS7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs, less than 60% with recombinant rat IL-1 Rrp2 and recombinant human IL-1 Rrp2 is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-1 R4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title