Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-2, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF21212250U
Encompass_Preferred
Click to view available options
Quantity:
250 μg
5 μg

Recombinant Protein

Recombinant Murine IL-2 is a 17.2 kDa protein containing 148 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04351
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 16183
Molecular Weight (g/mol) 17.2 kDa
Name Mouse IL-2, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 250 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Common Name IL-2
Gene Symbol IL2
Biological Activity Determined by its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence PTSSSTSSST AEAQQQQQQQ QQQQQHLEQL LMDLQELLSR MENYRNLKLP RMLTFKFYLP KQATELKDLQ CLEDELGPLR HVLDLTQSKS FQLEDAENFI SNIRVTVVKL KGSDNTFECQ FDDESATVVD FLRRWIAFCQ SIISTSPQ
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.