Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-2 R beta Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5891V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5891V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5891V100 Supplier R&D Systems Supplier No. FAB5891V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-2 R beta
Applications Flow Cytometry
Classification Monoclonal
Clone 130122
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P16297
Gene Alias CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Gene Symbols IL2RB
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3560
Test Specificity Detects mouse IL-2 R beta in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R alpha , rhIL-2 R beta , recombinant mouse (rm) IL-2 R gamma , rhIL-15 R alpha , or rmIL-21 R is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.