Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-20 R beta Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4388
This item is not returnable.
View return policy
Description
IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Mouse samples. It is validated for Western Blot.Specifications
IL-20 R beta/FNDC6 | |
Polyclonal | |
Biotin | |
NP_001032323 | |
IL20RB | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-20 R beta Asp14-Asn215 Accession # NP_001032323 | |
50 μg | |
Primary | |
Detects mouse IL-20 R beta in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human IL-20 R beta is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FNDC6, DIRS1, FNDC6, IL20R beta, IL-20R2, IL-20Rb, IL20RB, IL20RB interleukin 20 receptor beta, MGC34923 | |
Sheep | |
Affinity Purified | |
RUO | |
53833 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction