Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-21 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC594A025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC594A025 25 Tests
IC594A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. IC594A025 Supplier R&D Systems Supplier No. IC594A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 1 publication

IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-21
Applications Flow Cytometry
Classification Monoclonal
Clone 149204
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9ES17
Gene Alias CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11
Gene Symbols IL21
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59067
Test Specificity Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.