Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-21R Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF596
This item is not returnable.
View return policy
Description
IL-21R Polyclonal specifically detects IL-21R in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
IL-21R | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
Goat | |
Affinity Purified | |
RUO | |
50615 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
Q9JHX3 | |
IL21R | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-21 R Leu21-Pro236 Accession # Q9JHX3 | |
100 μg | |
Primary | |
Detects mouse IL-21 R in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human IL-21 R is observed, and less than 1% cross-reactivity will recombinant mouse IL-2 R beta is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction