Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-22 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC582T100UG
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-22 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9JJY9 | |
IL22 | |
E. coli-derived recombinant mouse IL-22 Leu34-Val179 Accession # Q9JJY9 | |
RUO | |
50616 | |
Mouse | |
IgG2a |
Flow Cytometry | |
140301 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Rat | |
100 μg | |
Primary | |
Detects mouse IL-22 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant rat IL-22 is observed, 15% cross-reactivity with recombinant human IL-22 is observed, and no cross-reactivity with recombinant mouse IL-10 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction