Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-22 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF582
This item is not returnable.
View return policy
Description
IL-22 Polyclonal specifically detects IL-22 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-22 | |
Polyclonal | |
Biotin | |
Q9JJY9 | |
IL22 | |
E. coli-derived recombinant mouse IL-22 Leu34-Val179 Accession # Q9JJY9 | |
50 μg | |
Primary | |
Detects mouse IL-22 in ELISAs and Western blots. In sandwich immunoassays, approximately 75% cross-reactivity with recombinant rat IL-22 is observed and less than 25% cross-reactivity with recombinant human IL-22 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Goat | |
Affinity Purified | |
RUO | |
50616 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction