Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-22 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC582P025UG
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC582P025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC582P025UG Supplier R&D Systems Supplier No. IC582P025UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 6 publications

IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-22
Applications Flow Cytometry
Classification Monoclonal
Clone 140301
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Accession No. Q9JJY9
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Gene Symbols IL22
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession No. Q9JJY9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 50616.0
Test Specificity Detects mouse IL-22 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant rat IL-22 is observed, 15% cross-reactivity with recombinant human IL-22 is observed, and no cross-reactivity with recombinant mouse IL-10 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.