Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-22 R alpha 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | IL-22R alpha 1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Western Blot.Specifications
IL-22R alpha 1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
IL22RA1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q80XZ4 | |
58985 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-22 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human IL-22 R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title