Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-22 R alpha 1 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB42941P
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-22R alpha 1 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
58985 | |
Mouse | |
Purified |
Flow Cytometry | |
496514 | |
Flow Cytometry 10 uL/10^6 cells | |
Q80XZ4 | |
IL22RA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4 | |
100 Tests | |
Primary | |
Detects mouse IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, 25% cross-reactivity with recombinant human (rh) IL-22 R alpha 1 is observed and no cross-reactivity with rhIL-20 R alpha , recombinant mouse (rm) IL-20 R alpha , rhIL-22BP, or rmIL-22BP is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction