Learn More
Mouse IL-22BP APC-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-22BP/IL22 RA2 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | AAI24999 |
| Gene Alias | class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 |
| Gene Symbols | IL22RA2 |
| Host Species | Sheep |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-22BP Thr19-Pro230 Accession # AAI24999 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.