Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-23 p19 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Supplier: R&D Systems AF1619
This item is not returnable.
View return policy
Description
IL-23A/IL-23 P19 Polyclonal specifically detects IL-23A/IL-23 P19 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IL-23A/IL-23 P19 | |
Polyclonal | |
Unconjugated | |
Q9EQ14 | |
IL23A | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-23 p19 Met1-Ala196 Accession # Q9EQ14 | |
50 μg | |
Primary | |
Detects mouse IL-23 p19 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse IL-12/23 p40 is observed. | |
Mouse | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.07-0.35 ug/mL | |
IL23, IL-23, IL-23 p19, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha | |
Goat | |
Affinity Purified | |
RUO | |
51561 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction