Learn More
Mouse IL-23R Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB16861U100UG
Catalog No. FB16861U100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-23R
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession # Q8K4B4
100 μg
149233
Mouse
Supernatant
Flow Cytometry
753317
Flow Cytometry 0.25-1 μg/106 cells
Q8K4B4
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects mouse IL-23 R in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.