Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-23R Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
$200.50 - $2020.00
Specifications
Antigen | IL-23R |
---|---|
Clone | 258010 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL |
Applications | Western Blot, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1686SP
![]() |
R&D Systems
MAB1686SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1686100
![]() |
R&D Systems
MAB1686100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1686500
![]() |
R&D Systems
MAB1686500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IL-23R | |
LYOPH | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
IL23R | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
258010 | |
Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q8K4B4 | |
149233 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-23 R in Western blots. In Western blots, this antibody does not cross-react with recombinant human IL-23 R. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title