Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-24 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | IL-24 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
IL-24 Polyclonal specifically detects IL-24 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IL-24 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
C49A, FISP, IL10B, IL24, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, MOB-5, ST16, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
IL24 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q925S4.1 | |
11009 | |
E. coli-derived recombinant mouse IL-24 Gln27-Leu181 Accession # Q925S4.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-24 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human IL-24 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title