Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-27 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | IL-27 |
---|---|
Clone | 355028 |
Concentration | LYOPH |
Dilution | Immunoprecipitation 25 ug/mL |
Applications | Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18343SP
![]() |
R&D Systems
MAB18343SP |
25 μg |
Each for $200.50
|
|
|||||
MAB18343
![]() |
R&D Systems
MAB18343 |
100 μg |
Each for $503.00
|
|
|||||
Description
IL-27 Monoclonal specifically detects IL-27 in Mouse samples. It is validated for Immunoprecipitation.Specifications
IL-27 | |
LYOPH | |
Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
O35228 (EBI3) & Q8K3I6 (p28) | |
246778 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28) Accession # O35228 (EBI3) & Q8K3I6 (p28) | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects mouse IL-27 heterodimer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-27 p28, rmIL-27 EBI-3, or recombinant human (rh) IL-27 heterodimer is observed. In Western blots, no cross-reactivity with rmIL-27 p28 or rhIL-27 heterodimer is observed. |
355028 | |
Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
IL27 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title