Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-27 R alpha/WSX-1/TCCR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB21091SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB21091SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB21091SP Supplier R&D Systems Supplier No. MAB21091SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 263503
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O70394
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Gene Symbols IL27RA
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 R alpha/WSX-1/TCCR Gly29-Lys510 Accession # O70394
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9466
Test Specificity Detects mouse IL-27 R alpha/WSX-1/TCCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse gp130 or recombinant human IL-27 R alpha is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.