Learn More
Mouse IL-27 R alpha/WSX-1/TCCR Fluorescein-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-27R alpha/WSX-1/TCCR |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 263503 |
| Conjugate | Fluorescein |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | O70394 |
| Gene Alias | class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 |
| Gene Symbols | IL27RA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.