Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-27 R alpha/WSX-1/TCCR PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB21091P100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB21091P100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB21091P100 Supplier R&D Systems Supplier No. FAB21091P100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Applications Flow Cytometry
Classification Monoclonal
Clone 263503
Conjugate R-PE
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O70394
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Gene Symbols IL27RA
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 R alpha/WSX-1/TCCR, Gly29-Lys510, Accession No. O70394
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9466.0
Test Specificity Detects mouse IL-27 R alpha/WSX-1/TCCR in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse gp130 or recombinant human IL-27 R alpha/WSX-1/TCCR is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.