Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-28A/B (IFN-lambda 2/3) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$200.50 - $2020.00
Specifications
Antigen | IL-28A/B (IFN-lambda 2/3) |
---|---|
Clone | 244716 |
Concentration | LYOPH |
Dilution | Neutralization 3-9 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17892SP
![]() |
R&D Systems
MAB17892SP |
25 μg |
Each for $200.50
|
|
|||||
MAB17892100
![]() |
R&D Systems
MAB17892100 |
100 μg |
Each for $573.00
|
|
|||||
MAB17892500
![]() |
R&D Systems
MAB17892500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
IL-28A/B (IFN-lambda 2/3) Monoclonal specifically detects IL-28A/B (IFN-lambda 2/3) in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
IL-28A/B (IFN-lambda 2/3) | |
LYOPH | |
Neutralization, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q8CGK6 | |
282616 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
244716 | |
Neutralization 3-9 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IFNl2, IFNlambda 2, IFN-lambda 2, IFN-lambda 3, IL28A, IL-28A, IL28B, interferon lambda 2 | |
E. coli-derived recombinant mouse IL-28B/IFN-lambda 3 Asp20-Val193 Accession # Q8CGK6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-28B/IFN-lambda 3 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant mouse IL-28A/IFN-lambda 2 and no cross-reactivity with recombinant human (rh) IL-28A or rhIL-29 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title