Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-28A/IFN-lambda 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4635
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4635 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4635 Supplier R&D Systems Supplier No. MAB4635
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Mouse samples. It is validated for Neutralization.

Specifications

Antigen IL-28A/IFN-lambda 2
Applications Neutralization
Classification Monoclonal
Clone 625616
Conjugate Unconjugated
Dilution Neutralization 2-10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001019844
Gene Alias interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2
Gene Symbols IFNL2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-28A/IFN-lambda 2 Asp20-Val193 Accession # NP_001019844
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282616
Test Specificity Detects mouse IL-28A/IFN-lambda 2 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse IL-28B and no cross-reactivity with recombinant human (rh) IL-28A, rhIL-28B, or rhIL-29 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.