Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-31 Antibody, R&D Systems™

Rat Monoclonal Antibody
$188.28 - $579.52
Specifications
| Antigen | IL-31 |
|---|---|
| Clone | 323407 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
IL-31 Monoclonal specifically detects IL-31 in Mouse samples. It is validated for Western Blot.Specifications
| IL-31 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q6EAL8 | |
| 386653 | |
| E. coli-derived recombinant mouse IL-31 Thr24-Cys163 Accession # Q6EAL8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-31 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-31. |
| 323407 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL31, IL-31interleukin-31, interleukin 31 | |
| IL31 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title