Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-31RA Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2107
This item is not returnable.
View return policy
Description
IL-31RA Polyclonal specifically detects IL-31RA in Mouse samples. It is validated for Western Blot.Specifications
IL-31RA | |
Polyclonal | |
Unconjugated | |
AAS82848 | |
IL31RA | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-31 RA Met28-Glu531 Accession # AAS82848 | |
100 μg | |
Primary | |
Detects mouse IL-31 RA in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
Goat | |
Affinity Purified | |
RUO | |
133396 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction