Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-33 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3626G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3626G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3626G100 Supplier R&D Systems Supplier No. IC3626G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-33 Monoclonal specifically detects IL-33 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-33
Applications Flow Cytometry
Classification Monoclonal
Clone 396118
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.5 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8BVZ5
Gene Alias C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Gene Symbols IL33
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-33 Ser109-Ile266 Accession # Q8BVZ5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90865
Test Specificity Detects mouse IL-33 in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.