Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-33 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 16 publications
Supplier: R&D Systems MAB3626SP
This item is not returnable.
View return policy
Description
IL-33 Monoclonal specifically detects IL-33 in Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
IL-33 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Q8BVZ5 | |
IL33 | |
E. coli-derived recombinant mouse IL-33 Ser109-Ile266 Accession # Q8BVZ5 | |
25 μg | |
Primary | |
Detects mouse IL-33 in direct ELISAs. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
396118 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
90865 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction