Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2297
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2297 100 μg
AF2297SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2297 Supplier R&D Systems Supplier No. AF2297
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

IL-36 alpha/IL-1F6 Polyclonal specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen IL-36 alpha/IL-1F6
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction 40 ug/mL
Gene Accession No. Q9JLA2
Gene Alias FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha
Gene Symbols Il1f6
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-36 alpha/IL-1F6 Met1-His160 Accession # Q9JLA2
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27179
Test Specificity Detects mouse IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) IL-1F5, rhIL-1F6, and rhIL-1F9 is observed, and less than 2% cross-reactivity with recombinant mouse IL-1F8, rhIL-1F7, rhIL-1 alpha , rhIL-1 beta , rhIL-1ra, and rhIL-18 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.