Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-36 beta/IL-1F8 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | IL-36 beta/IL-1F8 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
IL-36 beta/IL-1F8 Polyclonal specifically detects IL-36 beta/IL-1F8 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
IL-36 beta/IL-1F8 | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Mouse | |
family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, IL36 beta, IL36B, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta | |
Il1f8 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q9D6Z6 | |
27177 | |
E. coli-derived recombinant mouse IL-36 beta/IL-1F8 Met1-Lys183 Accession # Q9D6Z6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-36 beta/IL-1F8 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) IL-1F8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title