Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-36Ra/IL-1F5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2714
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2714 Supplier R&D Systems Supplier No. MAB2714
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-36Ra/IL-1F5 Monoclonal specifically detects IL-36Ra/IL-1F5 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IL-36Ra/IL-1F5
Applications Western Blot
Classification Monoclonal
Clone 759207
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9QYY1
Gene Alias FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5, IL1F5 (Canonical product IL-1F5a), IL-1HY1, IL-1L1, IL-1RP3, IL36RA, IL-36RA, IL36RN, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist
Gene Symbols IL36RN
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-36Ra/IL-1F5 Met1-Cys155 Accession # Q9QYY1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26525
Test Specificity Detects mouse IL-36Ra/IL-1F5 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human IL-36Ra and recombinant mouse IL-1ra is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.