Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-36RA Recombinant Protein, PeproTech®

Catalog No. p-7246201
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine IL-36RA is an E.coli-derived 16.9 kDa protein containing 154 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9QYY1
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 54450
Molecular Weight (g/mol) 16.9 kDa
Name Mouse IL-36 RA
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI413231; FIL1; FIL1 delta; FIL1(DELTA); FIL1D; Fil1delta; IL-1 delta; IL-1 related protein 3; Il1f5; IL-1F5; IL1F5 (Canonical product IL-1F5a); IL-1F5 (IL-1 HY1, FIL1-delta, IL-1 RP3, IL-1L1, IL-1-delta); Il1h3; Il-1h3; IL1HY1; IL-1 HY1; IL1L1; IL-1L1; IL-1 ra homolog 1; IL-1-related protein 3; IL1RP3; IL-1 RP3; IL36RA; IL-36 Ra; IL36RN; Interleukin; interleukin 1 family, member 5; interleukin 1 family, member 5 (delta); interleukin 1 receptor antagonist homolog 1; interleukin 36 receptor antagonist; interleukin-1 delta; Interleukin-1 family member 5; interleukin-1 family member 5; interleukin-36 receptor antagonist protein; interleukin-1 homolog 3; Interleukin-1 HY1; interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; interleukin-36 receptor antagonist protein; MGC29840; PSORP; PSORS14; UNQ1896/PRO4342
Common Name IL1F5
Gene Symbol Il1f5
Biological Activity Measured by its ability to inhibit secretion of IL-8 by A431 cells in the presence of human IL-36 gamma.
Conjugate Unconjugated
Recombinant Recombinant
Sequence VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGV QGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.