Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-38/IL-1F10 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems IC7774G
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-38/IL-1F10 | |
Monoclonal | |
Alexa Fluor 488 | |
Q8R459 | |
IL1F10 | |
E. coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession # Q8R459 | |
100 Tests | |
84639 | |
Mouse | |
Purified |
Flow Cytometry | |
798036 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse IL-38/IL-1F10 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction