Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-3R beta Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5492U100UG
This item is not returnable.
View return policy
Description
IL-3R beta Monoclonal specifically detects IL-3R beta in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-3R beta | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P26954 | |
Csf2rb2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta His23-Trp440 Accession # P26954 | |
RUO | |
1439 | |
Mouse | |
IgG2a |
Flow Cytometry | |
130705 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3R beta, IL-3R beta, IL-3Rb, IL3RB, Il3rb2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse IL-3 R beta in direct ELISAs. Cross-reactivity with AIC2B was not tested. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction