Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-4R alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$200.50 - $667.00
Specifications
| Antigen | IL-4R alpha |
|---|---|
| Clone | 129801 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Mouse samples. It is validated for Western Blot.Specifications
| IL-4R alpha | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| Il4ra | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 129801 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P16382 | |
| 3566 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-4 R alpha Ile26-Arg233 Accession # P16382 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-4 R alpha in direct ELISAs and Western blots. Shows less than 1% cross-reactivity with recombinant human IL-4 R alpha . |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title