Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF405SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF405SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF405SP Supplier R&D Systems Supplier No. AF405SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-5 Polyclonal specifically detects IL-5 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-5
Applications Western Blot, Neutralization, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.4-0.9 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Gene Symbols IL5
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-5
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Test Specificity Detects mouse IL-5 in direct ELISAs and Western blots. Neutralizes the biological activity of recombinant mouse IL-5, but will not neutralize the biological activity of recombinant human IL-5. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human IL-5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.