Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-5 R alpha/CD125 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | IL-5R alpha/CD125 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-5R alpha/CD125 Polyclonal specifically detects IL-5R alpha/CD125 in Mouse samples. It is validated for Western Blot.Specifications
IL-5R alpha/CD125 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
IL5RA | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P21183 | |
3568 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-5 R alpha/CD125 Asp18-His339 Accession # P21183 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-5 R alpha/CD125 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IL-5 R alpha is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and rhIL-13 R alpha 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title