Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse IL-6 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit (2 Plates)
1 Pk. (6 Plates)
Sufficient For:
2 x 96-well Plates
6 x 96-well Plates
Description
The Mouse IL-6 Quantikine ELISA Kit quantifies mouse IL-6 in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).
Specifications
Specifications
Assay Range | 7.80 to 500 pg/mL |
Assay Sensitivity | 1.8 pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (Heparin), Plasma (Heparin), Serum, Serum |
Specificity | Natural and Recombinant Mouse IL-6 |
Sufficient For | 6 x 96-well Plates |
Target Species | Mouse |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction