Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB4072SP
This item is not returnable.
View return policy
Description
IL-7 Monoclonal specifically detects IL-7 in Mouse samples. It is validated for Neutralization.Specifications
IL-7 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
E. coli-derived recombinant mouse IL-7, Ser543-Thr870, Accession # Q99814 | |
25 μg | |
3574 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Neutralization | |
1005101 | |
Neutralization 75-450 ng/mL | |
Q99814 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse IL-7 in direct ELISAs. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction