Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-7 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | IL-7 |
---|---|
Clone | 2304B |
Dilution | Neutralization 0.5-3 μg/mL |
Applications | Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4071SP
![]() |
R&D Systems
MAB4071SP |
25 μg |
Each for $200.50
|
|
|||||
MAB4071100
![]() |
R&D Systems
MAB4071100 |
100 μg |
Each for $479.50
|
|
|||||
Description
IL-7 Monoclonal specifically detects IL-7 in Mouse samples. It is validated for Neutralization.Specifications
IL-7 | |
Neutralization 0.5-3 μg/mL | |
Monoclonal | |
Supernatant | |
Mouse | |
P10168 | |
3574 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2304B | |
Neutralization | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
E. coli-derived recombinant mouse IL-7, Glu26-Ile154, Accession # P10168 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-7 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title