Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-7 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF407
This item is not returnable.
View return policy
Description
IL-7 Polyclonal specifically detects IL-7 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-7 | |
Polyclonal | |
Biotin | |
Q544C8 | |
IL7 | |
E. coli-derived recombinant mouse IL-7 (R&D Systems, Catalog # 407-ML) Glu26-Ile154 Accession # Q544C8 | |
50 μg | |
Primary | |
Detects mouse IL-7 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with rhIL-7 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
Goat | |
Affinity Purified | |
RUO | |
3574 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction