Learn More
Mouse IL-7R alpha/CD127 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Shop All R&D Systems Products


Description
Specifications
Specifications
| Antigen | IL-7R alpha/CD127 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1140A |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P16872 |
| Gene Alias | CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha |
| Host Species | Rabbit |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.