Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-7R alpha/CD127 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $561.50
Specifications
Antigen | IL-7R alpha/CD127 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
IL-7R alpha/CD127 Polyclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IL-7R alpha/CD127 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
IL7R | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q9R0C1 | |
3575 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-7 R alpha/CD127 Glu21-Asp239 Accession # Q9R0C1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-7 R alpha in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human IL-7 R alpha is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title